Gene Transcripts

Gene Symbol
ATG16L1
Entrez Gene ID
55054
Gene Name
autophagy related 16 like 1
Gene Aliases
APG16L, ATG16A, ATG16L, IBD10, WDR30
Chromosome Location
Chr. 2: 233251571 - 233295674 on Build GRCh38
UniGene ID
Hs.529322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190266.1 NP_001177195.1 9 1182
NM_001190267.1 NP_001177196.1 9 1180
NM_017974.3 NP_060444.3 8 1127
NM_030803.6 NP_110430.5 9 1184
NM_198890.2 NP_942593.2 5 695
XM_005246082.2 XP_005246139.1 10 1172
XM_005246084.2 XP_005246141.1 7 711
XM_005246086.2 XP_005246143.1 6 660
XM_006712608.2 XP_006712671.1 9 983
GenBank mRNA AK000897.1 - 8 795
AK001690.1 - 8 973
AK024453.1 - 8 1722
AK027854.1 - 7 709
AK074434.1 - 3 154
AK123876.1 - 6 651
AK308400.1 - 8 1025
AY358182.1 - 9 1073
AY398617.1 - 9 1073
BC071846.1 - 5 533
BC117337.1 - 9 846
BC143900.1 - 8 789
EF079889.1 - 8 870
EF079890.1 - 5 438
EF560734.1 - 8 997
JQ924062.1 - 8 870

View Full Product Details