Gene Transcripts

Gene Symbol
LRRC20
Entrez Gene ID
55222
Gene Name
leucine rich repeat containing 20
Gene Aliases
Chromosome Location
Chr. 10: 70298970 - 70382650 on Build GRCh38
UniGene ID
Hs.7778
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278211.1 NP_001265140.1 5 735
NM_001278212.1 NP_001265141.1 5 680
NM_001278213.1 NP_001265142.1 4 479
NM_001278214.1 NP_001265143.1 4 535
NM_018205.3 NP_060675.1 4 512
NM_018239.3 NP_060709.2 4 530
NM_207119.2 NP_997002.1 5 993
NR_103467.1 3 385
NR_103468.1 3 367
XM_017016374.1 XP_016871863.1 5 675
XM_017016375.1 XP_016871864.1 5 670
GenBank mRNA AK001613.1 - 4 488
AK001706.1 - 4 506
AL122117.1 - 2 217
AY358318.1 - 5 737
BC024001.2 - 5 671 4822655
CF552977.1 - 5 649
DA032832.1 - 3 359
DA160782.1 - 4 511
DA386385.1 - 3 341
DA704994.1 - 4 479

View Full Product Details