Gene Transcripts

Gene Symbol
WTAP
Entrez Gene ID
9589
Gene Name
Wilms tumor 1 associated protein
Gene Aliases
Mum2
Chromosome Location
Chr. 6: 159726680 - 159756319 on Build GRCh38
UniGene ID
Hs.446091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270531.1 NP_001257460.1 5 389
NM_001270532.1 NP_001257461.1 5 521
NM_001270533.1 NP_001257462.1 5 389
NM_004906.4 NP_004897.2 5 521
NM_152857.2 NP_690596.1 5 389
NM_152858.2 NP_690597.1 5 521
XM_017011514.1 XP_016867003.1 5 359
XM_017011515.1 XP_016867004.1 3 621
XM_017011516.1 XP_016867005.1 5 359
GenBank mRNA AF374416.1 - 5 402
AJ276706.1 - 4 169
AK127822.1 - 5 400
AK290854.1 - 5 402
BC000383.2 - 5 380
BC004432.1 - 5 380
BC069192.1 - 5 357 4653831
BE906617.1 - 5 506
BG702882.1 - 5 393
BQ948913.1 - 5 327
BX387163.2 - 5 371
CN284477.1 - 5 351
D14661.1 - 5 301
DA138013.1 - 5 521
DA561768.1 - 5 395

View Full Product Details