Gene Transcripts
- Gene Symbol
- SORBS2
- Entrez Gene ID
- 8470
- Gene Name
- sorbin and SH3 domain containing 2
- Gene Aliases
- ARGBP2, PRO0618
- Chromosome Location
- Chr. 4: 185585444 - 185956716 on Build GRCh38
- UniGene ID
- Hs.481342
- Species
- Homo sapiens
View Full Product Details