Gene Transcripts
- Gene Symbol
- NARF
- Entrez Gene ID
- 26502
- Gene Name
- nuclear prelamin A recognition factor
- Gene Aliases
- IOP2
- Chromosome Location
- Chr. 17: 82458184 - 82490537 on Build GRCh38
- UniGene ID
- Hs.256526
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001038618.2 |
NP_001033707.1
|
3, 4 |
558 |
|
NM_012336.3 |
NP_036468.1
|
3, 4 |
400 |
|
NM_031968.2 |
NP_114174.1
|
3, 4 |
400 |
|
XM_005256340.3 |
XP_005256397.1
|
3, 4 |
566 |
|
XM_006722278.3 |
XP_006722341.1
|
3, 4 |
488 |
|
XM_011523541.2 |
XP_011521843.1
|
3, 4 |
490 |
|
XM_011523543.2 |
XP_011521845.1
|
3, 4 |
306 |
|
XM_011523544.2 |
XP_011521846.1
|
3, 4 |
497 |
|
XM_017024449.1 |
XP_016879938.1
|
3, 4 |
543 |
|
XM_017024450.1 |
XP_016879939.1
|
3, 4 |
543 |
|
GenBank mRNA |
AF128406.1 |
- |
3, 4 |
351 |
|
AK000929.1 |
- |
3, 4 |
353 |
|
AK056966.1 |
- |
3, 4 |
533 |
|
AK297875.1 |
- |
3, 4 |
400 |
|
AK303155.1 |
- |
3, 4 |
353 |
|
BC000438.2 |
- |
3, 4 |
367 |
2820621 |
View Full Product Details