Gene Transcripts

Gene Symbol
NDUFAF7
Entrez Gene ID
55471
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 7
Gene Aliases
C2orf56, MidA, PRO1853
Chromosome Location
Chr. 2: 37231631 - 37259052 on Build GRCh38
UniGene ID
Hs.433466
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144736.4 NP_653337.1 6, 7 764
XM_005264410.1 XP_005264467.1 6, 7 744
XM_011532956.1 XP_011531258.1 6, 7 700
GenBank mRNA AK302150.1 - 5, 6 634
BC004548.2 - 6, 7 729 3959139
BC012374.1 - 1, 2 78
BI334629.1 - 6, 7 561

View Full Product Details