Gene Transcripts

Gene Symbol
HS1BP3
Entrez Gene ID
64342
Gene Name
HCLS1 binding protein 3
Gene Aliases
ETM2, HS1-BP3
Chromosome Location
Chr. 2: 20553861 - 20651661 on Build GRCh38
UniGene ID
Hs.531785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022460.3 NP_071905.3 7 1916
XM_017004700.1 XP_016860189.1 7 2015
GenBank mRNA AK024311.1 - 6 1328
BC050636.1 - 7 1896 5193161

View Full Product Details