Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199771.1 | NP_001186700.1 | 2 | 276 | |
NM_002905.3 | NP_002896.2 | 2 | 271 | ||
GenBank mRNA | AK293313.1 | - | 2 | 227 | |
BI765016.1 | - | 2 | 182 | ||
BP381700.1 | - | 2 | 271 | ||
U43559.1 | - | 2 | 164 | ||
U89717.1 | - | 2 | 201 |