Gene Transcripts

Gene Symbol
CLEC3B
Entrez Gene ID
7123
Gene Name
C-type lectin domain family 3 member B
Gene Aliases
TN, TNA
Chromosome Location
Chr. 3: 45026207 - 45036073 on Build GRCh38
UniGene ID
Hs.476092
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003278.2 NP_003269.2 1 182
XM_017007116.1 XP_016862605.1 1 242
GenBank mRNA AK312093.1 - 1 190
BC011024.1 - 1 147 4042921
BI837909.1 - 1 266
CD616729.1 - 1 156
CR541981.1 - 1 86
CR542009.1 - 1 86
X64559.1 - 1 179

View Full Product Details