Gene Transcripts

Gene Symbol
TRABD
Entrez Gene ID
80305
Gene Name
TraB domain containing
Gene Aliases
LP6054, PP2447
Chromosome Location
Chr. 22: 50185889 - 50199599 on Build GRCh38
UniGene ID
Hs.517731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320484.1 NP_001307413.1 8 836
NM_001320485.1 NP_001307414.1 8 824
NM_001320487.1 NP_001307416.1 8 858
NM_001320488.1 NP_001307417.1 8 858
NM_025204.3 NP_079480.2 8 849
NR_135275.1 8 820
XM_006724422.3 XP_006724485.1 8 828
XM_006724423.3 XP_006724486.1 8 821
XM_011530712.2 XP_011529014.2 8 896
XM_011530713.2 XP_011529015.2 8 894
XM_011530715.2 XP_011529017.2 8 899
XM_011530716.2 XP_011529018.1 8 859
XM_011530717.2 XP_011529019.1 8 840
XM_011530718.2 XP_011529020.1 8 819
XM_017028943.1 XP_016884432.1 8 901
XM_017028944.1 XP_016884433.1 8 936
GenBank mRNA AF218001.1 - 1 984
AK024447.1 - 1 3768
AK292050.1 - 8 828
AL449244.1 - 8 830
AY203945.1 - 8 816
BC012445.1 - 8 668 3857552
BC093029.1 - 8 843 30527560
BX389277.2 - 2 116
BX417817.2 - 8 809
BX425224.2 - 8 803

View Full Product Details