Gene Transcripts

Gene Symbol
CYB5B
Entrez Gene ID
80777
Gene Name
cytochrome b5 type B
Gene Aliases
CYB5-M, CYPB5M, OMB5
Chromosome Location
Chr. 16: 69424595 - 69466264 on Build GRCh38
UniGene ID
Hs.461131
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030579.2 NP_085056.2 2 284
GenBank mRNA AB009282.1 - 2 311
AK291576.1 - 2 216
BC004373.1 - 2 219 3639133
BC014431.1 - 2 242 3542294
BX647922.1 - 2 260

View Full Product Details