Gene Transcripts

Gene Symbol
KRTAP3-1
Entrez Gene ID
83896
Gene Name
keratin associated protein 3-1
Gene Aliases
KAP3.1, KRTAP3.1
Chromosome Location
Chr. 17: 41008521 - 41009114 on Build GRCh38
UniGene ID
Hs.307027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031958.1 NP_114164.1 1 411
GenBank mRNA AJ406931.1 - 1 411

View Full Product Details