Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270420.1 | NP_001257349.1 | 3 | 379 | |
NM_001270421.1 | NP_001257350.1 | 3 | 379 | ||
NM_032545.3 | NP_115934.1 | 3 | 379 | ||
XM_011511486.2 | XP_011509788.1 | 3 | 374 | ||
GenBank mRNA | AF312769.1 | - | 3 | 137 | |
AK290094.1 | - | 3 | 355 | ||
AK315326.1 | - | 3 | 379 | ||
BC069508.1 | - | 3 | 161 | 7262189 | |
BC074825.2 | - | 3 | 140 | 30915323 | |
BC074826.2 | - | 3 | 140 | 30915540 | |
BC110080.1 | - | 3 | 252 | 40030641 | |
BC146897.1 | - | 3 | 256 | 9056825 | |
BM509218.1 | - | 3 | 313 | ||
CA777249.1 | - | 3 | 352 | ||
CA941621.1 | - | 3 | 311 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079530.1 | NP_001072998.1 | 3 | 482 | |
XM_005263737.4 | XP_005263794.1 | 3 | 364 | ||
GenBank mRNA | BQ777283.1 | - | 3 | 324 | |
BU784123.1 | - | 3 | 337 |