Gene Transcripts

Gene Symbol
TM2D3
Entrez Gene ID
80213
Gene Name
TM2 domain containing 3
Gene Aliases
BLP2
Chromosome Location
Chr. 15: 101632977 - 101652391 on Build GRCh38
UniGene ID
Hs.288912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307960.1 NP_001294889.1 2 161
NM_001308026.1 NP_001294955.1 3 239
NM_025141.3 NP_079417.2 2 161
NM_078474.2 NP_510883.2 3 239
GenBank mRNA AF353992.1 - 2 136
AK026257.1 - 2 151
AK094955.1 - 1 571
AK296553.1 - 3 230
AK297048.1 - 2 146
AK315584.1 - 3 215
BC006150.1 - 3 211 3051281
BC008873.2 - 2 134 4137334
BU595412.1 - 3 234

View Full Product Details