Gene Transcripts
- Gene Symbol
- STK32C
- Entrez Gene ID
- 282974
- Gene Name
- serine/threonine kinase 32C
- Gene Aliases
- PKE, YANK3
- Chromosome Location
- Chr. 10: 132207482 - 132332559 on Build GRCh38
- UniGene ID
- Hs.469002
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318878.1 |
NP_001305807.1
|
4, 5 |
816 |
|
NM_001318879.1 |
NP_001305808.1
|
4, 5 |
677 |
|
NM_173575.3 |
NP_775846.2
|
4, 5 |
778 |
|
NR_134911.1 |
|
4, 5 |
455 |
|
XM_011539688.1 |
XP_011537990.1
|
4, 5 |
849 |
|
XM_011539690.2 |
XP_011537992.1
|
4, 5 |
764 |
|
XM_011539693.2 |
XP_011537995.1
|
4, 5 |
782 |
|
XM_011539694.1 |
XP_011537996.1
|
4, 5 |
516 |
|
XM_011539695.1 |
XP_011537997.1
|
4, 5 |
717 |
|
XM_011539696.1 |
XP_011537998.1
|
4, 5 |
524 |
|
GenBank mRNA |
AK057849.1 |
- |
4, 5 |
455 |
|
AK098366.1 |
- |
3, 4 |
615 |
|
AK302102.1 |
- |
4, 5 |
763 |
|
AK308956.1 |
- |
4, 5 |
855 |
|
AK310725.1 |
- |
4, 5 |
782 |
|
AY098866.1 |
- |
4, 5 |
778 |
|
BC045760.1 |
- |
4, 5 |
667 |
4831007 |
View Full Product Details