Gene Transcripts

Gene Symbol
CHMP4B
Entrez Gene ID
128866
Gene Name
charged multivesicular body protein 4B
Gene Aliases
C20orf178, CHMP4A, CTPP3, CTRCT31, SNF7, SNF7-2, Shax1, VPS32B, Vps32-2, dJ553F4.4
Chromosome Location
Chr. 20: 33811304 - 33854367 on Build GRCh38
UniGene ID
Hs.472471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176812.4 NP_789782.1 1 305
GenBank mRNA AB100261.1 - 1 250
AY329085.1 - 1 140
AY726594.1 - 1 151
BC033859.1 - 1 249 5173394
BQ926070.1 - 299
CT001913.1 - 1 305

View Full Product Details