Gene Transcripts

Gene Symbol
SSTR1
Entrez Gene ID
6751
Gene Name
somatostatin receptor 1
Gene Aliases
SRIF-2, SS-1-R, SS1-R, SS1R
Chromosome Location
Chr. 14: 38207999 - 38213063 on Build GRCh38
UniGene ID
Hs.248160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001049.2 NP_001040.1 3 1056
GenBank mRNA BC035618.1 - 1 713
BX248780.1 - 3 1056

View Full Product Details