Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001169109.1 | NP_001162580.1 | 2 | 473 | |
NM_001169110.1 | NP_001162581.1 | 2 | 424 | ||
NM_001169111.1 | NP_001162582.1 | 2 | 442 | ||
NM_005138.2 | NP_005129.2 | 2 | 413 | ||
GenBank mRNA | AL021683.1 | - | 2 | 351 | |
BC102024.1 | - | 2 | 341 | 40030584 | |
BC102025.1 | - | 2 | 341 | 40030589 | |
BG392981.1 | - | 2 | 424 | ||
BM787182.1 | - | 2 | 472 | ||
BQ878572.1 | - | 2 | 413 | ||
CN409692.1 | - | 2 | 381 | ||
CR456569.1 | - | 1 | 271 |