Gene Transcripts

Gene Symbol
GNG12
Entrez Gene ID
55970
Gene Name
G protein subunit gamma 12
Gene Aliases
Chromosome Location
Chr. 1: 67701466 - 67833779 on Build GRCh38
UniGene ID
Hs.431101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018841.5 NP_061329.3 3 278
XM_017001809.1 XP_016857298.1 3 445
XM_017001810.1 XP_016857299.1 4 686
XM_017001811.1 XP_016857300.1 4 615
GenBank mRNA AF119663.1 - 3 262
AF123766.1 - 3 269
AF365871.1 - 3 205
AF493879.1 - 1 73
AK223000.1 - 3 220
AK293101.1 - 4 219
AL832431.1 - 3 231
BC005940.1 - 3 259 4049838
BP236225.1 - 3 265
BP236699.1 - 3 278

View Full Product Details