Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019888.3 | NP_063941.3 | 1 | 106 | |
GenBank mRNA | AY227893.1 | - | 1 | 105 | |
BC069105.1 | - | 1 | 106 | 7216921 | |
BC069599.1 | - | 1 | 105 | 7262270 | |
BC096702.1 | - | 1 | 105 | 40016185 | |
BC096737.1 | - | 1 | 105 | 40016189 | |
BC098169.1 | - | 1 | 105 | 40016186 | |
BC098351.1 | - | 1 | 105 | 40016184 |