Gene Transcripts

Gene Symbol
FKRP
Entrez Gene ID
79147
Gene Name
fukutin related protein
Gene Aliases
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5
Chromosome Location
Chr. 19: 46746015 - 46758575 on Build GRCh38
UniGene ID
Hs.515493
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039885.2 NP_001034974.1 4 522
NM_024301.4 NP_077277.1 4 470
XM_005259247.2 XP_005259304.1 5 583
XM_005259248.2 XP_005259305.1 5 573
XM_005259249.4 XP_005259306.1 5 515
XM_011527306.1 XP_011525608.1 3 319
XM_011527307.1 XP_011525609.1 4 368
XM_017027297.1 XP_016882786.1 4 689
GenBank mRNA AJ314847.1 - 4 458
AK022638.1 - 4 458
AK095497.1 - 4 509
AK291282.1 - 4 458
BC002612.2 - 2 266 3160297
DA123579.1 - 4 399
DN999716.1 - 1 191

View Full Product Details