Gene Transcripts
- Gene Symbol
- Ascl1
- Entrez Gene ID
- 64186
- Gene Name
- achaete-scute family bHLH transcription factor 1
- Gene Aliases
- Mash1
- Chromosome Location
- Chr. 7: 28038662 - 28040504 on Build Rnor_6.0
- UniGene ID
- Rn.32936
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022384.1 |
NP_071779.1
|
1 |
1135 |
|
GenBank mRNA |
X53725.1 |
- |
1 |
1135 |
|
View Full Product Details