Gene Transcripts
- Gene Symbol
- Kcnb1
- Entrez Gene ID
- 25736
- Gene Name
- potassium channel, voltage gated Shab-related subfamily B, member 1
- Gene Aliases
- DRK1PC, Kcr1-1, Kv2.1, Shab
- Chromosome Location
- Chr. 3: 163850785 - 163935610 on Build Rnor_6.0
- UniGene ID
- Rn.26724
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013186.1 |
NP_037318.1
|
1 |
254 |
|
GenBank mRNA |
X16476.1 |
- |
1 |
254 |
|
View Full Product Details