Gene Transcripts

Gene Symbol
Coil
Entrez Gene ID
50998
Gene Name
coilin
Gene Aliases
Chromosome Location
Chr. 10: 76320676 - 76336169 on Build Rnor_6.0
UniGene ID
Rn.203468
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017360.1 NP_059056.1 1, 2 253
GenBank mRNA AF220424.1 - 1, 2 253
AF399643.1 - 1, 2 253

View Full Product Details