Gene Transcripts

Gene Symbol
Slc15a3
Entrez Gene ID
246239
Gene Name
solute carrier family 15 (oligopeptide transporter), member 3
Gene Aliases
Pht2
Chromosome Location
Chr. 1: 226897853 - 226912644 on Build Rnor_6.0
UniGene ID
Rn.17317
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139341.1 NP_647557.1 1 270
GenBank mRNA AB026665.1 - 1 270

View Full Product Details