Gene Transcripts

Gene Symbol
Vipr1
Entrez Gene ID
24875
Gene Name
vasoactive intestinal peptide receptor 1
Gene Aliases
RATVASREC, VASREC
Chromosome Location
Chr. 8: 130283453 - 130313431 on Build Rnor_6.0
UniGene ID
Rn.9973
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012685.2 NP_036817.1 7 1204
GenBank mRNA BC087136.1 - 7 1204
M86835.1 - 7 1173

View Full Product Details