Gene Transcripts

Gene Symbol
Cnr1
Entrez Gene ID
25248
Gene Name
cannabinoid receptor 1 (brain)
Gene Aliases
SKR6R
Chromosome Location
Chr. 5: 49309324 - 49333064 on Build Rnor_6.0
UniGene ID
Rn.89774
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012784.4 NP_036916.1 2 433
XM_006237984.2 XP_006238046.1 2 585
GenBank mRNA U40395.1 - 1 281
X55812.1 - 2 433

View Full Product Details