Gene Transcripts

Gene Symbol
Mgmt
Entrez Gene ID
25332
Gene Name
O-6-methylguanine-DNA methyltransferase
Gene Aliases
Chromosome Location
Chr. 1: 209237255 - 209464189 on Build Rnor_6.0
UniGene ID
Rn.9836
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012861.1 NP_036993.1 4 384
GenBank mRNA M76704.1 - 3 329
S61804.1 - 4 384
X54862.1 - 4 410

View Full Product Details