Gene Transcripts

Gene Symbol
Tshr
Entrez Gene ID
25360
Gene Name
thyroid stimulating hormone receptor
Gene Aliases
TSHRA
Chromosome Location
Chr. 6: 115170287 - 115306871 on Build Rnor_6.0
UniGene ID
Rn.87913
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012888.1 NP_037020.1 2, 3 301
XM_008764742.1 XP_008762964.1 2, 3 873
GenBank mRNA M34842.1 - 2, 3 301

View Full Product Details