Gene Transcripts

Gene Symbol
Lipe
Entrez Gene ID
25330
Gene Name
lipase, hormone sensitive
Gene Aliases
HSL
Chromosome Location
Chr. 1: 82248031 - 82266727 on Build Rnor_6.0
UniGene ID
Rn.10566
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012859.1 NP_036991.1 1 573
GenBank mRNA BC078888.1 - 1 556
U40001.1 - 1 573

View Full Product Details