Gene Transcripts

Gene Symbol
Cd81
Entrez Gene ID
25621
Gene Name
Cd81 molecule
Gene Aliases
Tapa1
Chromosome Location
Chr. 1: 216237697 - 216253311 on Build Rnor_6.0
UniGene ID
Rn.1975
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013087.1 NP_037219.1 2 306
GenBank mRNA BC060583.1 - 2 137
FQ231991.1 - 2 292
FQ232886.1 - 2 294
U19894.1 - 2 306

View Full Product Details