Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017060.2 | NP_058756.2 | 4 | 254 | |
XM_006230687.2 | XP_006230749.1 | 3 | 1123 | ||
GenBank mRNA | AB298805.1 | - | 2 | 173 | |
CB613004.1 | - | 4 | 254 | ||
FQ224126.1 | - | 4 | 314 |