Gene Transcripts

Gene Symbol
Cartpt
Entrez Gene ID
29131
Gene Name
CART prepropeptide
Gene Aliases
Cart
Chromosome Location
Chr. 2: 30125249 - 30127693 on Build Rnor_6.0
UniGene ID
Rn.89164
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017110.1 NP_058806.1 2 240
XM_006231843.2 XP_006231905.1 2 625
GenBank mRNA U10071.1 - 2 240

View Full Product Details