Gene Transcripts

Gene Symbol
Slco1a1
Entrez Gene ID
50572
Gene Name
solute carrier organic anion transporter family, member 1a1
Gene Aliases
OATP-1, Oatp1, Slc21a1, Slc21a3
Chromosome Location
Chr. 4: 176158174 - 176231331 on Build Rnor_6.0
UniGene ID
Rn.231253
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017111.1 NP_058807.1 3 203
GenBank mRNA FQ209854.1 - 3 221
FQ219332.1 - 3 265
L19031.1 - 3 203

View Full Product Details