Gene Transcripts

Gene Symbol
Rps17
Entrez Gene ID
29286
Gene Name
ribosomal protein S17
Gene Aliases
Chromosome Location
Chr. 1: 143167329 - 143169915 on Build Rnor_6.0
UniGene ID
Rn.6920
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017152.3 NP_058848.2 3 267
GenBank mRNA BC058484.1 - 3 271
FQ209589.1 - 3 264
FQ210096.1 - 3 257
FQ221123.1 - 3 262
FQ221680.1 - 3 257
FQ222308.1 - 3 257
FQ222382.1 - 3 269
FQ222858.1 - 3 262
FQ223398.1 - 3 264
FQ224175.1 - 3 257
FQ228331.1 - 3 262
FQ228517.1 - 3 264
FQ229910.1 - 3 264
K02933.1 - 3 265

View Full Product Details