Gene Transcripts
- Gene Symbol
- Btnl1
- Entrez Gene ID
- 100038862
- Gene Name
- butyrophilin-like 1
- Gene Aliases
- Btnl3, Gm316, Gm33
- Chromosome Location
- Chr. 17: 34377123 - 34386162 on Build GRCm38
- UniGene ID
- Mm.440426
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC052925.1 |
- |
9 |
3499 |
|
View Full Product Details