Gene Transcripts

Gene Symbol
Esp3
Entrez Gene ID
100126766
Gene Name
exocrine gland secreted peptide 3
Gene Aliases
Gm21945
Chromosome Location
Chr. 17: 40632075 - 40637060 on Build GRCm38
UniGene ID
Mm.471591
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251916.1 NP_001238845.1 3 797
GenBank mRNA AB306981.1 - 3 797

View Full Product Details