Gene Transcripts

Gene Symbol
Btbd18
Entrez Gene ID
100270744
Gene Name
BTB (POZ) domain containing 18
Gene Aliases
Btbd18-ps, Gm13718, OTTMUSG00000013494
Chromosome Location
Chr. 2: 84659072 - 84668837 on Build GRCm38
UniGene ID
Mm.393636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145100.1 NP_001138572.1 5 1171

View Full Product Details