Gene Transcripts

Gene Symbol
Gm17660
Entrez Gene ID
100271704
Gene Name
predicted gene, 17660
Gene Aliases
Scpppq1
Chromosome Location
Chr. 5: 104070060 - 104077608 on Build GRCm38
UniGene ID
Mm.366664
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163772.1 NP_001157244.1 9 346
GenBank mRNA EU642618.1 - 9 346
HM015622.1 - 9 374
HM015623.1 - 9 330

View Full Product Details