Gene Transcripts

Gene Symbol
Fam50b
Entrez Gene ID
108161
Gene Name
family with sequence similarity 50, member B
Gene Aliases
D0H6S2654E, X5L, XAP-5-like
Chromosome Location
Chr. 13: 34734828 - 34747623 on Build GRCm38
UniGene ID
Mm.41991
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138746.2 NP_620085.2 2 1278
GenBank mRNA AK016195.1 - 2 1181
BC049659.1 - 2 1191
Y18506.1 - 2 1179

View Full Product Details