Gene Transcripts
- Gene Symbol
- Hist2h3c1
- Entrez Gene ID
- 15077
- Gene Name
- histone cluster 2, H3c1
- Gene Aliases
- BE691662, H3-615, H3f2, Hist2h3ca1
- Chromosome Location
- Chr. 3: 96246685 - 96247348 on Build GRCm38
- UniGene ID
- Mm.422680
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC094041.1 |
- |
|
1491 |
|
View Full Product Details