Gene Transcripts

Gene Symbol
Eme2
Entrez Gene ID
193838
Gene Name
essential meiotic endonuclease 1 homolog 2 (S. pombe)
Gene Aliases
2810013J18Rik
Chromosome Location
Chr. 17: 24892152 - 24895087 on Build GRCm38
UniGene ID
Mm.440199
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163102.1 NP_001156574.1 8 1227
GenBank mRNA AK012738.1 - 7 1287
BC034637.1 - 1266
BC092228.1 - 8 1185

View Full Product Details