Gene Transcripts

Gene Symbol
Clint1
Entrez Gene ID
216705
Gene Name
clathrin interactor 1
Gene Aliases
AI642036, AW537616, C530049I24Rik, Enth, Epn4, F630206G17Rik, mKIAA0171
Chromosome Location
Chr. 11: 45851930 - 45910625 on Build GRCm38
UniGene ID
Mm.169673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045520.3 NP_001038985.2 3 390
GenBank mRNA AB093212.1 - 2 1477
AK143544.1 - 3 391
AK147875.1 - 3 375
AK168991.1 - 3 356

View Full Product Details