Gene Transcripts

Gene Symbol
Prss38
Entrez Gene ID
216797
Gene Name
protease, serine 38
Gene Aliases
Gm249, Mpn2
Chromosome Location
Chr. 11: 59372669 - 59376253 on Build GRCm38
UniGene ID
Mm.339586
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045521.1 NP_001038986.1 1 116
GenBank mRNA AK145810.1 - 1 121
BC147400.1 - 1 112 9056455
BC147401.1 - 1 112 9056447
BY736706.1 - 1 116

View Full Product Details