Gene Transcripts

Gene Symbol
Fam208a
Entrez Gene ID
218850
Gene Name
family with sequence similarity 208, member A
Gene Aliases
4732418E12, 4933409E02Rik, D14Abb1e
Chromosome Location
Chr. 14: 27426658 - 27483555 on Build GRCm38
UniGene ID
Mm.247767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114879.1 NP_001108351.1 10 1406
NM_028945.1 NP_083221.1 10 1406
GenBank mRNA AK016751.1 - 10 1420
AK044563.1 - 10 1406

View Full Product Details