Gene Transcripts

Gene Symbol
Klhl42
Entrez Gene ID
232539
Gene Name
kelch-like 42
Gene Aliases
A330079N23Rik, AI450355, C230080I20Rik, Ctb9, Klhdc5, mKIAA1340
Chromosome Location
Chr. 6: 147091075 - 147112778 on Build GRCm38
UniGene ID
Mm.491206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081237.1 NP_001074706.1 3 5061
GenBank mRNA AK129335.1 - 3 3971
BC011326.1 - 1 2554
BC030345.1 - 1 2565

View Full Product Details