Gene Transcripts

Gene Symbol
Zbtb34
Entrez Gene ID
241311
Gene Name
zinc finger and BTB domain containing 34
Gene Aliases
Gm349, mKIAA1993
Chromosome Location
Chr. 2: 33406108 - 33431324 on Build GRCm38
UniGene ID
Mm.296512
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085507.1 NP_001078976.1 3 3093
NM_001276332.1 NP_001263261.1 2 2973
GenBank mRNA AK173321.1 - 2 2966

View Full Product Details