Gene Transcripts

Gene Symbol
Hdx
Entrez Gene ID
245596
Gene Name
highly divergent homeobox
Gene Aliases
D030011N01Rik, Gm380
Chromosome Location
Chr. X: 111575204 - 111697079 on Build GRCm38
UniGene ID
Mm.246623
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080549.2 NP_001074018.1 3 1250
NM_001290459.1 NP_001277388.1 2 1103
GenBank mRNA AK083436.1 - 2 1091
BC116300.1 - 3 1193 40095063
BC116301.1 - 2 1046 40095064

View Full Product Details