Gene Transcripts

Gene Symbol
CMKLR1
Entrez Gene ID
1240
Gene Name
chemerin chemokine-like receptor 1
Gene Aliases
CHEMERINR, ChemR23, DEZ, RVER1
Chromosome Location
Chr. 12: 108288044 - 108339348 on Build GRCh38
UniGene ID
Hs.197143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142343.1 NP_001135815.1 4 1548
NM_001142344.1 NP_001135816.1 3 1335
NM_001142345.1 NP_001135817.1 3 1162
NM_004072.2 NP_004063.1 3 1472
XM_017018820.1 XP_016874309.1 2 1290
GenBank mRNA AB112777.1 - 1 965
AK291800.1 - 3 1329
AY497547.1 - 1 965
BC106927.2 - 4 1362 40033186
BC106928.1 - 4 1362 40033187
BT019556.1 - 1 965
BT019557.1 - 1 965
U79526.1 - 3 1325
U79527.1 - 3 1478

View Full Product Details