Gene Transcripts

Gene Symbol
Mphosph9
Entrez Gene ID
269702
Gene Name
M-phase phosphoprotein 9
Gene Aliases
4930548D04Rik, 9630025B04Rik, AW413446, AW547060, B930097C17Rik, C87456, MPP-9, MPP9
Chromosome Location
Chr. 5: 124250959 - 124330759 on Build GRCm38
UniGene ID
Mm.60224
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081323.2 NP_001074792.1 22 6831
NM_001277867.1 NP_001264796.1 23 6921
GenBank mRNA AK079326.1 - 1 1293

View Full Product Details